missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ CD84 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gly 225) of human CD84 (NP_003865.1) fused to a polyhistidine tag at the C-terminus

Manufacturer:  Life Technologies LS10100H08H50

 View more versions of this product

Catalog No. 10100H08H50


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; CD84 Recombinant Human Protein, His Tag; 50ug; Source: Human; Purity: greater than or equal to 90% by SDS-PAGE

Cluster of Differentiation 84 (CD84), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
8832
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Other Proteins
DKFZp781E2378, LY9B, SLAMF5, hCD84, mCD84.
CD84
CD Proteins
His Tag
Lyophilized
24kDa
50μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
CD84 molecule
Assay Standard
Mammalian