Please login to your online account to display your discounted pricing

Sino Biological™ CD80 (B7-1) Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Asn 242) of human CD80 precursor (NP_005182.1) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10698H08H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10698H08H50

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Family CD Proteins
Protein Tag His Tag
Common Name CD80 molecule
Validated Application Assay Standard
Molecular Weight 25.4kDa
Gene ID (Entrez) 941
Purity ≥90% by SDS-PAGE
Gene Alias CD28LG, CD28LG1, LAB7
Format Lyophilized
Expression System Mammalian
Structural Form Truncated/Recombinant/Cell Surface Protein/Ligand
Protein Subtype Other Proteins
Quantity 50μg
Gene Symbol CD80
Endotoxin Concentration <1 EU/μg
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories

Cluster of Differentiation 80 (CD80/B7-1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CD80-His at 1μg/well can bind human CD28-Fc with a linear range of 0.8-4.0μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.