missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD80 (B7-1) Recombinant Human Protein, hIgG1-Fc.His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Asn 242) of human CD80 precursor (NP_005182.1) precursor fused to the His-tagged Fc region of human IgG1 at the C-terminus

$305.00 - $830.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CD28LG, CD28LG1, LAB7
Gene ID (Entrez) 941
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10698H03H50 View Documents Life Technologies
LS10698H03H50
50μg Each for $305.00
 
10698H03HL View Documents Life Technologies
LS10698H03H250
5 x 50μg Each for $830.00
 
Description

Description

Cluster of Differentiation 80 (CD80/B7-1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CD28-His at 1μg/well can bind human CD80-Fch with a linear range of 1.25-40.0μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
941
<1 EU/μg
CD28LG, CD28LG1, LAB7

For Research Use Only. Not for use in diagnostic procedures.