missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ CD80 (B7-1) Recombinant Human Protein, hIgG1-Fc.His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Asn 242) of human CD80 precursor (NP_005182.1) precursor fused to the His-tagged Fc region of human IgG1 at the C-terminus

Manufacturer:  Life Technologies LS10698H03H50

 View more versions of this product

Catalog No. 10698H03H50


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; CD80 (B7-1) Recombinant Human Protein, hIgG1-Fc.His Tag; 50ug; Source: Human; Purity: greater than or equal to 90% by SDS-PAGE

Cluster of Differentiation 80 (CD80/B7-1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CD28-His at 1μg/well can bind human CD80-Fch with a linear range of 1.25-40.0μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
941
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Other Proteins
CD28LG, CD28LG1, LAB7
CD80
CD Proteins
His Tag/IgG1 Fc Tag
Lyophilized
51.7kDa
50μg
Truncated/Recombinant/Cell Surface Protein/Ligand
<1 EU/μg
CD80 molecule
Assay Standard
Mammalian