missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ CD69 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human CD69 (NP_001772.1) extracellular domain (Ser 62-Lys 199) fused to a signal peptide at the N-terminus and a polyhistidine tag at the C-terminus

$144.00 - $1,788.00

Specifications

Purity ≥85% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CLEC2C, CD69
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11150H08H25 Life Technologies
LS11150H08H25
5 x 5μg Each for $350.00
11150H08H5 Life Technologies
LS11150H08H5
5μg Each for $144.00
11150H08HL Life Technologies
LS11150H08H250
5 x 50μg Each for $1,788.00
Specifications

Description

CD69 Molecule (CD69), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥85% by SDS-PAGE
<1 EU/μg
CLEC2C, CD69
PAGE
Lyophilized
969