missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ CD63 Recombinant Human Protein, His Tag DFS Item

This recombinant human protein is expressed from a DNA sequence encoding the second the extracellular domain (Ala 103-Val 203) of human CD63 (P08962-1) fused to both a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus

Manufacturer:  Life Technologies LS11271H08H250

 View more versions of this product

Catalog No. 11271H08HL


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; CD63 Recombinant Human Protein, His Tag; 5 x 50ug; Source: Human; Purity: greater than or equal to 90% by SDS-PAGE

Cluster of Differentiation 63 (CD63) recombinant human protein is supplied as a lyophilized powder. This protein is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.&line-break;Background: CD63, is a multipass membrane protein which belongs to the tetraspanin (TM4SF) family. Tetraspanins participate in a variety of cellular processes, like cell activation, adhesion, differentiation and tumour invasion. CD63/Tspan-30 is expressed in melanoma and appears to be involved in regulation of motility and adhesion of melanoma cells. CD63/Tspan-30 is associated with early stages of melanoma tumor progression. It may play a role in growth regulation. Lack of expression of CD63 in platelets has been observed in a patient with Hermansky-Pudlak syndrome (HPS), which is a genetically heterogeneous, rare, autosomal recessive disorder characterized by oculocutaneous albinism, bleeding due to platelet storage pool deficiency, and lysosomal storage defects.
  • Activity: This recombinant protein has not been tested
  • Formulation: lyophilized in PBS, pH 7.4, 5% mannitol, 5% trehalose, and 0.02% Tween®-80
  • Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. Manufactured by Sino Biological (China).

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
967
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Glycoproteins
LAMP-3, ME491, MLA1, OMA81H, TSPAN30
CD63
CD Proteins
His Tag
Lyophilized
13kDa
5 x 50μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
CD63 molecule
Assay Standard
Mammalian