Please login to your online account to display your discounted pricing

Sino Biological™ CD5L Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human CD5L (NP_005885.1) (Met 1-Gly 347) fused to a C-terminal polyhistidine tag was expressed

Manufacturer: life technologies  LS10791H08H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10791H08H25

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 922
Endotoxin Concentration <1 EU/μg
Validated Application Assay Standard
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Expression System Mammalian
Purity ≥90% by SDS-PAGE
Protein Tag His Tag
Purification Method PAGE
Quantity 5 x 5μg
Common Name CD5 antigen-like
Protein Subtype Other Proteins
Protein Family CD Proteins
Structural Form Truncated/Recombinant
Termination C-Terminal
Gene Symbol CD5L
Molecular Weight 37.5kDa
Gene Alias UNQ203/PRO229, AIM, API6, PRO229, SP-ALPHA, Spalpha, CD5L

Cluster of Differentiation 5 Antigen-Like (CD5L), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 20mM Tris, 500mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.