Please login to your online account to display your discounted pricing

Sino Biological™ CD5 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human CD5 (NP_055022.2) (Met 1-Pro 372) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS11027H08H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11027H08H25

  • / Each
 

Description & Specifications

Specifications

Gene Alias LEU1, T1, CD5
Common Name CD5 Molecule
Protein Tag His Tag
Molecular Weight 40kDa
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Validated Application Assay Standard
Protein Family CD Proteins
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Protein Subtype Other Proteins
Gene Symbol CD5
Quantity 5 x 5μg
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Purity ≥90% by SDS-PAGE

CD5 molecule (CD5), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to bind biotinylated recombinant human LCK in a functional ELISA
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.