missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD48 Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding mature form of the mouse CD48 (NP_031675.1) (Met 1-Arg 216) fused to a polyhistidine tag at the C-terminus

$222.00 - $222.00

Specifications

Purity >85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CD48, AI449234, AW610730, BCM1, BLAST, BLAST-1, BLAST1, Bcm-1, MEM-102, SLAMF2, Sgp-60
View More Specs

Includes: Five vials of lyophilized protein

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50415M08H25 View Documents Life Technologies
LS50415M08H25
5 x 5μg Each for $222.00
 
Description

Description

CD48 antigen (CD48), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: 1. Measured by its ability to bind human 2B4 in a functional ELISA; 2. Measured by its ability to bind biotinylated recombinant mouse CD2 in a functional ELISA
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

>85% by SDS-PAGE
<1 EU/μg
Lyophilized
CD48, AI449234, AW610730, BCM1, BLAST, BLAST-1, BLAST1, Bcm-1, MEM-102, SLAMF2, Sgp-60

For Research Use Only. Not for use in diagnostic procedures.