Please login to your online account to display your discounted pricing

Sino Biological™ CD44 Recombinant Human Protein, His Tag DFS Item


This recombinant human protein is expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 220) of human CD44 (P16070-1) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS12211H08H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 12211H08H25

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Validated Application Assay Standard
Quantity 5 x 5μg
Gene ID (Entrez) 960
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Gene Symbol CD44
Expression System Mammalian
Structural Form Truncated/Recombinant/Cell Surface Protein
Protein Subtype Glycoproteins
Common Name CD44 molecule (Indian blood group)
Format Lyophilized
Molecular Weight 23.4kDa
Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Gene Alias AL133330.1, CDW44, CSPG8, ECMR-III, HCELL, HUTCH-I, IN, LHR, MC56, MDU2, MDU3, mgC10468
Protein Tag His Tag
Protein Family CD Proteins

Cluster of Differentiation 44 (CD44) recombinant human protein is supplied as a lyophilized powder. This protein is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.&line-break;Background: CD44, short for Cluster of Differentiation 44, is an adhesion molecule of the hyaluronate receptor family. The adhesive function of CD44, like other molecules involved in inducible adhesion, is tightly regulated. Posttranslational modifications, isoform expression, aggregation state, and protein associations all can affect the ligand binding properties of CD44, and these can vary depending on the cell type and the activation state of the cell. The principal ligand of CD44 is hyaluronan, and CD44 is involved in the metabolism of this compound. Furthermore, an increasing quantity of evidence suggests that CD44 has various functions related to inflammatory disease. CD44 deficiency induces severe liver injury. CD44-hyaluronate mediates in lymphocyte T and monocyte adhesion to the endothelium, stimulates proinflammatory cytokine release from macrophages and participates in dedifferentiation phenotype of smooth muscle cells from contractile state to synthetic one. CD44 acts in atherosclerotic plaque destabilization and neointimal proliferation after percutaneous vascular interventions. CD44 participates in many stages of atherosclerosis. CD44 also is a cell membrane protein often overexpressed on tumor cells and, being both a cell-cell and cell-extracellular matrix adhesion protein, is well positioned to contribute to this process.

  • N-terminal Sequence Analysis: Gln 21
  • Activity: This recombinant protein has not been tested
  • Formulation: lyophilized in PBS, pH 7.4, 5% mannitol, 5% trehalose, and 0.02% Tween®-80
  • Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. Manufactured by Sino Biological (China).