Please login to your online account to display your discounted pricing

Sino Biological™ CD40LG (CD154) Recombinant Human Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the soluble form of human CD40LG (NP_000065.1) (Glu 108-Leu 261) fused to an N-terminal Met

$93.00 - $696.00

Specifications

Purity ≥90% by SDS-PAGE
Format Frozen
Gene Alias RP3-527F8.3, CD154, CD40L, HIGM1, IGM, IMD3, T-BAM, TNFSF5, TRAP, gp39, hCD40L
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10239H08E25

DFS Item
 
life technologies
LS10239H08E25
5 x 5μg Each for $200.00

10239H08E5

DFS Item
 
life technologies
LS10239H08E5
5μg Each for $93.00

10239H08EL

DFS Item
 
life technologies
LS10239H08E250
5 x 50μg Each for $696.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Format Frozen
Gene Alias RP3-527F8.3, CD154, CD40L, HIGM1, IGM, IMD3, T-BAM, TNFSF5, TRAP, gp39, hCD40L

CD40 Ligand (CD40LG), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA: 1. Immobilized human CD40 at 2μg/mL (100μL/well) can bind biotinylated human CD40LG with a linear range of 15.6-500ng/mL; 2. Immobilized human CD40/Fc at 2μg/mL (100μL/well) can bind biotinylated human CD40LG with a linear range of 7.8-125ng/mL
  • Formulation: PBS, 0.2μg/μL BSA, 10% glycerol, pH 7.4
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.