Please login to your online account to display your discounted pricing

Sino Biological™ CD40LG (CD154) Recombinant Human Protein DFS Item


This recombinant protein was expressed from a DNA sequence encoding the soluble form of human CD40LG (NP_000065.1) (Glu 108-Leu 261) fused to an N-terminal Met

Manufacturer: life technologies  LS10239H08E250

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10239H08EL

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 17.7kDa
Quantity 5 x 50μg
Purity ≥90% by SDS-PAGE
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Expression System E. coli
Common Name CD40 ligand
Protein Tag His Tag
Gene Alias RP3-527F8.3, CD154, CD40L, HIGM1, IGM, IMD3, T-BAM, TNFSF5, TRAP, gp39, hCD40L
Protein Subtype TNF (Tumor Necrosis Factor)|Other Proteins
Storage Requirements Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Gene Symbol CD40LG
Validated Application Assay Standard
Format Frozen

CD40 Ligand (CD40LG), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA: 1. Immobilized human CD40 at 2μg/mL (100μL/well) can bind biotinylated human CD40LG with a linear range of 15.6-500ng/mL; 2. Immobilized human CD40/Fc at 2μg/mL (100μL/well) can bind biotinylated human CD40LG with a linear range of 7.8-125ng/mL
  • Formulation: PBS, 0.2μg/μL BSA, 10% glycerol, pH 7.4
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.