missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ CD40 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Arg 193) of human CD40 (NP_001241.1) fused to a C-terminal polyhistidine tag

$121.00 - $1,392.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CD40, Bp50, CDW40, mgC9013, TNFRSF5, p50
Gene ID (Entrez) 958
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10774H08H25 Life Technologies
LS10774H08H25
5 x 5μg Each for $241.00
10774H08H5 Life Technologies
LS10774H08H5
5μg Each for $121.00
10774H08H50 Life Technologies
LS10774H08H50
50μg Each for $383.00
10774H08HL Life Technologies
LS10774H08H250
5 x 50μg Each for $1,392.00
Specifications

Description

Cluster of Differentiation 40 (CD40), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human CD40 at 2μg/mL (100μL/well) can bind biotinylated human CD40LG with a linear range of 15.6-500ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
958
<1 EU/μg
CD40, Bp50, CDW40, mgC9013, TNFRSF5, p50