missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD4 Recombinant Human Protein, hIgG1-Fc.His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant human protein is expressed from a DNA sequence encoding the extracellular domain (Met 1-Trp 390) of human CD4 (NP_000607.1) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

$245.00 - $1,524.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CD4mut
Gene ID (Entrez) 920
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10400H03H25 View Documents Life Technologies
LS10400H03H25
5 x 5μg Each for $245.00
 
10400H03H50 View Documents Life Technologies
LS10400H03H50
50μg Each for $421.00
 
10400H03HL View Documents Life Technologies
LS10400H03H250
5 x 50μg Each for $1,524.00
 
Description

Description

Cluster of Differentiation 4 (CD4) recombinant human protein is supplied as a lyophilized powder. This protein is suitable for use in the study of protein-protein interactions, and cell biology research applications. It can also be used as a protein standard.
Background: Human T-cell surface glycoprotein CD4, also known as T-cell surface antigen T4/Leu-3, is a single-pass type I membrane protein. CD4 contains three Ig-like C2-type (immunoglobulin-like) domains and one Ig-like V-type (immunoglobulin-like) domain. CD4 is a glycoprotein expressed on the surface of T helper cells, regulatory T cells, monocytes, macrophages, and dendritic cells. CD4 amplifies the signal generated by the TCR. CD4 interacts directly with MHC class II molecules on the surface of the antigen presenting cell via its extracellular domain. CD4 is a primary receptor used by HIV-1 to gain entry into host T cells. HIV infection leads to a progressive reduction of the number of T cells possessing CD4 receptors. Therefore, medical professionals refer to the CD4 count to decide when to begin treatment for HIV-infected patients.
  • N-terminal Sequence Analysis: Lys26
  • Activity: measured by its binding ability in a functional ELISA; immobilized human CD4-His at 2μg/well has been shown to bind biotinylated HIV-1 GP120 with a linear range of 0.078-1.25μg/mL
  • Formulation: lyophilized in PBS, pH 7.4, 5% mannitol, 5% trehalose, and 0.02% Tween™-80
  • Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. Manufactured by Sino Biological (China).

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
920
<1 EU/μg
CD4mut

For Research Use Only. Not for use in diagnostic procedures.