Please login to your online account to display your discounted pricing

Sino Biological™ CD3E Recombinant Cynomolgus Protein, hIgG1-Fc Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the Cynomolgus (Macaca fascicularis) CD3E extracellular domain (Met 1 – Asp 117) fused to the Fc region of human IgG1 at the C-terminus

$2,070.00 - $2,070.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CD3E
View More Specs
Includes:  Five vials of lyophilized protein

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

90047C02HL

DFS Item
 
life technologies
LS90047C02H250
5 x 50μg Each for $2,070.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CD3E

CD3 Molecule, epsilon (CD3E), recombinant cynomplgus protein is supplied as a lyophilized powder.

  • Activity: This recombinant protein has not been tested
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.