missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ CD3D Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human CD3D extracellular domain (NP_000723.1) (Met 1-Ala 105) fused to a C-terminal polyhistidine tag

Manufacturer:  Life Technologies LS10981H08H50

 View more versions of this product

Catalog No. 10981H08H50


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; CD3D Recombinant Human Protein, His tag; 50ug; Source: Human; Purity: greater than or equal to 90% by SDS-PAGE

CD3D delta Molecule,(CD3D), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
915
PAGE
50μg
C-Terminal
<1 EU/μg
CD3D Molecule, delta (CD3-TCR complex)
Assay Standard
Mammalian
Lyophilized
11kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Truncated/Recombinant
CD3-DELTA, T3D, CD3D
CD3D
CD Proteins
His Tag