missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ CD3D & CD3E Recombinant Mouse Protein, hIgG1-Fc & hIgG1-Fc.His Tag DFS Item

CD3 Antigen delta and CD3 Antigen epsilon Heterodimer (CD3D & CD3E), recombinant mouse protein is supplied as a lyophilized powder

$397.00 - $397.00

Specifications

Purity ≥85% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias None
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Five vials of lyophilized protein


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CT025M2303H Life Technologies
LSCT025M2303H25
5μg Each for $397.00
Specifications

Description

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ala 105) of mouse CD3D (P04235) fused to the C-terminal flag-tagged Fc region of human IgG1 at the C-terminus, constructed the plasmid 1; a DNA sequence encoding the extracellular domain (Met 1-Asp 108) of mouse CD3E (P22646) fused to the C-terminal His-tagged Fc region of human IgG1 at the C-terminus, constructed the plasmid 2. The two plasmids were co-expressed and the mouse CD3D/CD3E heterodimer was purified.
  • Activity: This recombinant protein has not been tested
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥85% by SDS-PAGE
<1 EU/μg
None
PAGE
Lyophilized
12500/12501