missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD3D & CD3E Recombinant Human Protein, hIgG1-Fc & hIgG1-Fc.His Tag, Invitrogen™ Sino Biological™ DFS Item

CD3D molecule delta and CD3E molecule epsilon (CD3-TCR complex) Heterodimer (CD3D & CD3E), recombinant human protein is supplied as a lyophilized powder

$163.00 - $2,105.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias None
View More Specs

Includes: Five vials of lyophilized protein

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CT026H0323H View Documents Invitrogen
LSCT026H0323H5
5μg Each for $163.00
 
CT026H0323M View Documents Invitrogen
LSCT026H0323H25
5 x 5μg Each for $414.00
 
CT026H0323L View Documents Invitrogen
LSCT026H0323H250
5 x 50μg Each for $2,105.00
 
Description

Description

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ala 105) of human CD3D (P04234) fused to the C-terminal flag-tagged Fc region of human IgG1 at the C-terminus, constructed the plasmid 1; a DNA sequence encoding the extracellular domain (Met 1-Asp 126) of human CD3E (P07766) fused to the C-terminal His-tagged Fc region of human IgG1 at the C-terminus, constructed the plasmid 2. The two plasmids were co-expressed and the human CD3D/CD3E heterodimer was purified.
  • Activity: This recombinant protein has not been tested
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
None
PAGE
Lyophilized
915/916

For Research Use Only. Not for use in diagnostic procedures.