missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ CD38 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human CD38 (NP_001766.2) (Val 43-Ile 300) fused to a C-terminal polyhistidine tag

Manufacturer:  Life Technologies LS10818H08H50

 View more versions of this product

Catalog No. 10818H08H50


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; CD38 Recombinant Human Protein, His tag; 50ug; Source: Human; Purity: greater than or equal to 90% by SDS-PAGE

ADP-Ribosyl Cyclase (CD38), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
952
PAGE
50μg
C-Terminal
<1 EU/μg
CD38 Molecule
Assay Standard
Mammalian
Lyophilized
31.3kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
T10, CD38
CD38
CD Proteins
His Tag