Please login to your online account to display your discounted pricing

Sino Biological™ CD36 Recombinant Mouse Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of mouse CD36 (NP_001153030.1) (Gly 30-Lys 439) fused to a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus

$219.00 - $219.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias Cd36, FAT, GPIV, Scarb3
View More Specs
Includes:  Five vials of lyophilized protein

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50422M08H25

DFS Item
 
life technologies
LS50422M08H25
5 x 5μg Each for $219.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias Cd36, FAT, GPIV, Scarb3

CD36 Antigen (CD36), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human RSPO1 (Mfr. No. 11083-H08H) at 10μg/mL (100μL/well) can bind mouse CD36; the EC50 of mouse CD36 is 0.1-0.4μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.