Please login to your online account to display your discounted pricing

Sino Biological™ CD36 Recombinant Human Protein, His Tag DFS Item


This recombinant protein is expressed from a DNA sequence encoding the extracellular domain (Gly 30-Asn 439) of human CD36 (NP_001001547.1) fused both to a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus

Manufacturer: life technologies  LS10752H08H250

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10752H08HL

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Expression System Mammalian
Validated Application Assay Standard
Format Lyophilized
Gene ID (Entrez) 948
Quantity 5 x 50μg
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Purity ≥85% by SDS-PAGE
Gene Symbol CD36
Protein Tag His Tag
Protein Family CD Proteins
Endotoxin Concentration <1 EU/μg
Molecular Weight 48kDa
Structural Form Truncated/Recombinant/Cell Surface Protein
Protein Subtype Glycoproteins
Gene Alias CD36, CHDS7, FAT, GP3B, GP4, GPIV, PASIV, SCARB3
Common Name CD36 molecule (thrombospondin receptor)

Cluster of Differentiation 36 (CD36) recombinant human protein is supplied as a lyophilized powder. This protein is suitable for use in analysis of protein structure the study of protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.&line-break;Background: CD36, is integral membrane protein found on the surface of many cell types in vertebrate animals. It belongs to the CD36 family and is found on platelets, erythrocytes, monocytes, differentiated adipocytes, mammary epithelial cells, spleen cells and some skin microdermal endothelial cells. CD36 has also been implicated in hemostasis, thrombosis, malaria, inflammation, lipid metabolism and atherogenesis. CD36 seems to have numerous potential physiological functions. It binds to collagen, thrombospondin, anionic phospholipids and oxidized LDL. It directly mediates cytoadherence of Plasmodium falciparum parasitized erythrocytes and binds long chain fatty acids. CD36 may also function as a cell adhesion molecule and in the transport and/or as a regulator of fatty acid transport. Defects in CD36 are the cause of platelet glycoprotein IV deficiency also known as CD36 deficiency. As a receptor for thrombospondin 1, CD36 plays a role in the regulation of angiogenesis, which may be a therapeutic strategy for controlling the dissemination of malignant neoplasms.

  • N-terminal Sequence Analysis: Asp 31
  • Activity: measured by its binding ability in a functional ELISA; immobilized human CD36 at 20μg/mL (100μL/well) has been shown to bind biotinylated human RSPO1 with a linear range of 0.032-4μg/mL
  • Formulation: lyophilized in PBS, pH 7.4, 5% mannitol, 5% trehalose, and 0.02% Tween®-80
  • Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. Manufactured by Sino Biological (China).