Please login to your online account to display your discounted pricing

Sino Biological™ CD34 Recombinant Mouse Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of mouse CD34 (NP_001104529.1) (Met 1-Thr 287) fused to a polyhistidine tag at the C-terminus

$136.00 - $1,688.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias AU040960, Cd34
View More Specs
Includes:  Five vials of lyophilized protein

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50589M08H25

DFS Item
 
life technologies
LS50589M08H25
5 x 5μg Each for $338.00

50589M08H5

DFS Item
 
life technologies
LS50589M08H5
5μg Each for $136.00

50589M08H50

DFS Item
 
life technologies
LS50589M08H50
50μg Each for $538.00

50589M08HL

DFS Item
 
life technologies
LS50589M08H250
5 x 50μg Each for $1,688.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias AU040960, Cd34

CD34 Antigen (CD34), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

  • Activity: Measured by the ability of the immobilized protein to support the adhesion of the HUVEC human umbilical vein endothelial cell line; when 4 x 104 cells/well are added to mouse CD34 coated plates (0.8μg/mL, 100μL/well), approximately >30% will adhere after one hour at 37°C
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.