missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD34 Recombinant Human Protein, hIgG1-Fc Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of the human CD34 precursor (NP_001020280.1) (Met 1-Thr 290) fused to the Fc region of human IgG1 at the C-terminus

$460.00 - $460.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias RP11-328D5.2, CD34
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10103H02H50 View Documents Invitrogen
LS10103H02H50
50μg Each for $460.00
 
Description

Description

Cluster of Differentiation 34 (CD34), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in performing cell based assays. It can also be used as a protein standard, or in other research applications.
  • Activity: Measured by the ability of the immobilized protein to support the adhesion of the HUVEC human umbilical vein endothelial cell line; when 4 x 104 cells/well are added to human CD34 coated plates (100μL/well of a 0.8μg/mL solution), >40 % will adhere after one hour at 37°C
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Lyophilized
RP11-328D5.2, CD34

For Research Use Only. Not for use in diagnostic procedures.