missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ CD300A Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human CD300A (Q9UGN4-1) extracellular domain (Met 1-Gln 178) fused to a polyhistidine tag at the C-terminus

$217.00 - $217.00

Specifications

Purity ≥85% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CLM-8, CMRF-35-H9, CMRF-35H, CMRF35-H, CMRF35-H9, IGSF12, IRC1, IRC1/IRC2, IRC2, IRp60
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
12449H08H25 Life Technologies
LS12449H08H25
5 x 5μg Each for $217.00
Specifications

Description

Cluster of Differentiation 300A (CD300A), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥85% by SDS-PAGE
<1 EU/μg
CLM-8, CMRF-35-H9, CMRF-35H, CMRF35-H, CMRF35-H9, IGSF12, IRC1, IRC1/IRC2, IRC2, IRp60
PAGE
Lyophilized
11314