Please login to your online account to display your discounted pricing

Sino Biological™ CD300A Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human CD300A (Q9UGN4-1) extracellular domain (Met 1-Gln 178) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS12449H08H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 12449H08H25

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 18.8kDa
Purification Method PAGE
Purity ≥85% by SDS-PAGE
Quantity 5 x 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Termination C-Terminal
Structural Form Truncated/Recombinant
Protein Subtype Other Proteins
Endotoxin Concentration <1 EU/μg
Gene Alias CLM-8, CMRF-35-H9, CMRF-35H, CMRF35-H, CMRF35-H9, IGSF12, IRC1, IRC1/IRC2, IRC2, IRp60
Common Name CD300A Molecule
Gene Symbol CD300A
Protein Tag His Tag
Validated Application Assay Standard
Protein Family CD Proteins
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 11314

Cluster of Differentiation 300A (CD300A), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.