missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD28 Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Lys 149) of mouse CD28 (NP_031668.3) fused to a polyhistidine tag at the C-terminus

$82.00 - $522.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CD28, CH29-189K17.3
View More Specs

Includes: Five vials of protein in suspension buffer

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50103M08H5 View Documents Life Technologies
LS50103M08H5
5μg Each for $82.00
 
50103M08HL View Documents Life Technologies
LS50103M08H250
5 x 50μg Each for $522.00
 
Description

Description

Cluster of Differentiation 28 (CD28), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
CD28, CH29-189K17.3
PAGE
Lyophilized
12487

For Research Use Only. Not for use in diagnostic procedures.