missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD276 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Thr 461) of human CD276 (Q5ZPR3-1) was expressed fused to a polyhistidine tag at the C-terminus

$256.00 - $434.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CD276, PSEC0249, 4Ig-B7-H3, B7-H3, B7H3
Gene ID (Entrez) 80381
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11188H08H25 View Documents Life Technologies
LS11188H08H25
5 x 5μg Each for $256.00
 
11188H08H50 View Documents Life Technologies
LS11188H08H50
50μg Each for $434.00
 
Description

Description

Cluster of Differentiation 276 (CD276), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
80381
<1 EU/μg
CD276, PSEC0249, 4Ig-B7-H3, B7-H3, B7H3

For Research Use Only. Not for use in diagnostic procedures.