missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD274 Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of mouse CD274 (NP_068693.1) (Met 1-Thr 238) fused to a polyhistidine tag at the C-terminus

$223.00 - $802.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CD274, B7-H1, PD-L1, Pdcd1l1, Pdcd1lg1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50010M08H25 View Documents Invitrogen
LS50010M08H25
5 x 5μg Each for $223.00
 
50010M08H50 View Documents Invitrogen
LS50010M08H50
50μg Each for $287.00
 
50010M08HL View Documents Invitrogen
LS50010M08H250
5 x 50μg Each for $802.00
 
Description

Description

CD274 antigen (CD274), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant mouse CD274 at 1μg/mL (100μL/well) can bind mouse PD1 with a linear range of 6.25-400ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Lyophilized
CD274, B7-H1, PD-L1, Pdcd1l1, Pdcd1lg1

For Research Use Only. Not for use in diagnostic procedures.