Please login to your online account to display your discounted pricing

Sino Biological™ CD274 Recombinant Mouse Protein, hIgG1-Fc.His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Thr 238) of mouse CD274 (NP_068693.1) fused to the C-terminal His-tagged Fc region of human IgG1 at the C-terminus

$266.00 - $752.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CD274, B7-H1, PD-L1, Pdcd1l1, Pdcd1lg1
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50010M03H50

DFS Item
 
life technologies
LS50010M03H50
50μg Each for $266.00

50010M03HL

DFS Item
 
life technologies
LS50010M03H250
5 x 50μg Each for $752.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CD274, B7-H1, PD-L1, Pdcd1l1, Pdcd1lg1

CD274 Antigen (CD274), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant mouse PD-1 at 10μg/mL (100μL/well) can bind mouse CD274 with a linear range of 3.2-400ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.