missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ CD27 Recombinant Mouse Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Arg 182) of mouse CD27 (NP_001028298.1) fused to a polyhistidine tag at the C-terminus

$120.00 - $1,390.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias S152, Tnfrsf7, Tp55
Gene ID (Entrez) 21940
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50110M08H25 Life Technologies
LS50110M08H25
5 x 5μg Each for $246.00
50110M08H5 Life Technologies
LS50110M08H5
5μg Each for $120.00
50110M08HL Life Technologies
LS50110M08H250
5 x 50μg Each for $1,390.00
Specifications

Description

Cluster of Differentiation 27 (CD27), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant mouse CD27 at 2μg/mL (100μL/well) can bind human CD70 with a linear range of 78-1250pg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
21940
<1 EU/μg
S152, Tnfrsf7, Tp55