missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD244 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Arg 221) of human CD244 (NP_057466.1) fused to a polyhistidine tag at the C-terminus

$218.00 - $388.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias 2B4, CD244, NAIL, nMrk, NKR2B4, SLAMF4
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10042H08H25 View Documents Life Technologies
LS10042H08H25
5 x 5μg Each for $218.00
 
10042H08H50 View Documents Life Technologies
LS10042H08H50
50μg Each for $388.00
 
Description

Description

Cluster of Differentiation 244 (CD244), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CD244 at 2μg/mL (100μg/well) can bind human CD48, the EC50 of human CD48 is 0.39μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Lyophilized
2B4, CD244, NAIL, nMrk, NKR2B4, SLAMF4

For Research Use Only. Not for use in diagnostic procedures.