Please login to your online account to display your discounted pricing

Sino Biological™ CD24 Recombinant Human Protein, hIgG1-Fc Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human CD24 (NP_037362.1) without the pro peptide (Met 1-Gly 59) fused to the Fc region of human IgG1 at the C-terminus

Manufacturer: life technologies  LS11030H02H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11030H02H50

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 100133941
Protein Tag IgG1 Fc Tag
Gene Alias CD24A, FLJ22950, FLJ43543, mgC75043
Common Name CD 24 molecule
Quantity 50μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Expression System Mammalian
Termination C-Terminal
Endotoxin Concentration <1 EU/μg
Purity ≥90% by SDS-PAGE
Purification Method PAGE
Format Lyophilized
Structural Form Full Length/Recombinant
Gene Symbol CD24
Molecular Weight 30kDa
Protein Family CD Proteins
Validated Application Assay Standard

Cluster of Differentiation 24 (CD24), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.