missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD226 Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of mouse CD226 (NP_848802.2) (Met 1-Pro 254) fused to a polyhistidine tag at the C-terminus

$400.00 - $400.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias BC051526, DNAM-1, DNAM1, Pta1, TLiSA1, Cd226
View More Specs

Includes: Five vials of lyophilized protein

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50232M08H50 View Documents Invitrogen
LS50232M08H50
50μg Each for $400.00
 
Description

Description

CD226 antigen (CD226), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized mouse CD226 at 2μg/mL (100μL/well) can bind mouse PVR/CD155 with a linear range of 6.4-160ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Lyophilized
BC051526, DNAM-1, DNAM1, Pta1, TLiSA1, Cd226

For Research Use Only. Not for use in diagnostic procedures.