missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD226 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human CD226 (NP_006557.2) extracellular domain (Met 1-Asn 247) fused to a polyhistidine tag at the C-terminus

$222.00 - $222.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CD226, DNAM-1, DNAM1, PTA1, TLiSA1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10565H08H25 View Documents Invitrogen
LS10565H08H25
5 x 5μg Each for $222.00
 
Description

Description

CD226 molecule (CD226), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CD226 at 2μg/mL (100μL/well) can bind human PVRL2/CD112 with a linear range of 6.4-800ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Lyophilized
CD226, DNAM-1, DNAM1, PTA1, TLiSA1

For Research Use Only. Not for use in diagnostic procedures.