Please login to your online account to display your discounted pricing

Sino Biological™ CD226 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human CD226 (NP_006557.2) extracellular domain (Met 1-Asn 247) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10565H08H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10565H08H25

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 40kDa
Protein Subtype Glycoproteins|Intercellular Adhesion Molecule
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Format Lyophilized
Quantity 5 x 5μg
Purity ≥90% by SDS-PAGE
Gene Symbol DNAM-1/CD226
Protein Family CD Proteins
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Validated Application Assay Standard
Endotoxin Concentration <1 EU/μg
Protein Tag His Tag
Gene Alias CD226, DNAM-1, DNAM1, PTA1, TLiSA1
Common Name CD226 Molecule
Expression System Mammalian

CD226 molecule (CD226), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CD226 at 2μg/mL (100μL/well) can bind human PVRL2/CD112 with a linear range of 6.4-800ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.