missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD22 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Arg 687) of human CD22-β (P20273-1) fused to a polyhistidine tag at the C-terminus

$95.50 - $429.00

Specifications

Purity ≥95% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias RP11-446H13.1, FLJ41758, FLJ43475, FLJ44957
Gene ID (Entrez) 933
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11958H08H5 View Documents Life Technologies
LS11958H08H5
5μg Each for $95.50
 
11958H08H50 View Documents Life Technologies
LS11958H08H50
50μg Each for $429.00
 
Description

Description

Cluster of Differentiation 22 (CD22), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥95% by SDS-PAGE
Lyophilized
933
<1 EU/μg
RP11-446H13.1, FLJ41758, FLJ43475, FLJ44957

For Research Use Only. Not for use in diagnostic procedures.