missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD209 Recombinant Human Protein, hIgG1-Fc Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the N-terminally truncated extracellular (Lys 62-Ala 404) domain of human CD209 (NP_066978.1) fused to the Fc region of human IgG1 at the N-terminus

$435.00 - $435.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CDSIGN, CLEC4L, DC-SIGN, DC-SIGN1, mgC129965, CD209
Gene ID (Entrez) 30835
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10200H01H50 View Documents Invitrogen
LS10200H01H50
50μg Each for $435.00
 
Description

Description

Cluster of Differentiation 209 (CD209), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
30835
<1 EU/μg
CDSIGN, CLEC4L, DC-SIGN, DC-SIGN1, mgC129965, CD209

For Research Use Only. Not for use in diagnostic procedures.