Please login to your online account to display your discounted pricing

Sino Biological™ CD209 Recombinant Human Protein, hIgG1-Fc Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the N-terminally truncated extracellular (Lys 62-Ala 404) domain of human CD209 (NP_066978.1) fused to the Fc region of human IgG1 at the N-terminus

Manufacturer: life technologies  LS10200H01H250

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10200H01HL

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 65.8kDa
Gene ID (Entrez) 30835
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Common Name CD209 molecule
Gene Alias CDSIGN, CLEC4L, DC-SIGN, DC-SIGN1, mgC129965, CD209
Gene Symbol CD209
Structural Form Truncated/Recombinant/Cell Surface Protein
Endotoxin Concentration <1 EU/μg
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Format Lyophilized
Expression System Mammalian
Protein Tag IgG1 Fc Tag
Validated Application Assay Standard
Protein Subtype Lectin Receptor
Purity ≥90% by SDS-PAGE
Protein Family CD Proteins
Quantity 5 x 50μg

Cluster of Differentiation 209 (CD209), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.