missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD200R1 Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 238) of mouse CD200R1 (NP_067300.1) fused to a C-terminal polyhistidine tag at the C-terminus

$253.00 - $398.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CD200R, Mox2r, OX2R
Gene ID (Entrez) 57781
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50209M08H25 View Documents Life Technologies
LS50209M08H25
5 x 5μg Each for $253.00
 
50209M08H50 View Documents Life Technologies
LS50209M08H50
50μg Each for $398.00
 
Description

Description

Cluster of Differentiation 200 receptor 1 (CD200R1), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.
  • Activity: Measured by its ability to bind mouse CD200 in a functional ELISA; immobilized mouse CD200-His at 0.2μg/well can bind mouse CD200R1-His with a linear range of 0.8-4.0μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥85% by SDS-PAGE
Lyophilized
57781
<1 EU/μg
CD200R, Mox2r, OX2R

For Research Use Only. Not for use in diagnostic procedures.