missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ CD200R1 Recombinant Mouse Protein, hIgG1-Fc Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 238) of mouse CD200R1 precursor (NP_067300.1) fused to the Fc region of human IgG1 at the C-terminus

$246.00 - $1,390.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CD200R, Mox2r, OX2R
Gene ID (Entrez) 57781
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50209M03H25 Life Technologies
LS50209M03H25
5 x 5μg Each for $246.00
50209M03H50 Life Technologies
LS50209M03H50
50μg Each for $391.00
50209M03HL Life Technologies
LS50209M03H250
5 x 50μg Each for $1,390.00
Specifications

Description

Cluster of Differentiation 200 receptor 1 (CD200R1), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant mouse CD200 at 1μg/mL (100μL/well) can bind mouse CD200R1-Fc with a linear range of 1.5-200ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
57781
<1 EU/μg
CD200R, Mox2r, OX2R