missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD200 Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gly 232) of mouse CD200 (NP_001076017.1) fused to a polyhistidine tag at the C-terminus

$125.00 - $1,442.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CD200, Mox2, OX2
Gene ID (Entrez) 17470
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50074M08H5 View Documents Life Technologies
LS50074M08H5
5μg Each for $125.00
 
50074M08H50 View Documents Life Technologies
LS50074M08H50
50μg Each for $397.00
 
50074M08HL View Documents Life Technologies
LS50074M08H250
5 x 50μg Each for $1,442.00
 
Description

Description

Cluster of Differentiation 200 (CD200), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant mouse CD200 at 1μg/mL (100μL/well) can bind mouse CD200R1-Fc with a linear range of 1.5-200ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
17470
<1 EU/μg
CD200, Mox2, OX2

For Research Use Only. Not for use in diagnostic procedures.