missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD200 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gly 232) of human CD200 (NP_005935.4) fused to a polyhistidine tag at the C-terminus

$127.00 - $1,202.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CD200, MOX1, MOX2, MRC, OX-2
Gene ID (Entrez) 4345
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10886H08H5 View Documents Invitrogen
LS10886H08H5
5μg Each for $127.00
 
10886H08H25 View Documents Invitrogen
LS10886H08H25
5 x 5μg Each for $247.00
 
10886H08HL View Documents Invitrogen
LS10886H08H250
5 x 50μg Each for $1,202.00
 
Description

Description

Cluster of Differentiation 200 (CD200), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human CD200 at 1μg/mL (100μL/well) can bind human CD200R1-Fc with a linear range of 0.12-16ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
4345
<1 EU/μg
CD200, MOX1, MOX2, MRC, OX-2

For Research Use Only. Not for use in diagnostic procedures.