Please login to your online account to display your discounted pricing

Sino Biological™ CD200 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gly 232) of human CD200 (NP_005935.4) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10886H08H250

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10886H08HL

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 24kDa
Purity ≥90% by SDS-PAGE
Quantity 5 x 50μg
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Structural Form Truncated/Recombinant/Cell Surface Protein
Protein Subtype Glycoproteins
Endotoxin Concentration <1 EU/μg
Gene Alias CD200, MOX1, MOX2, MRC, OX-2
Common Name CD200 molecule
Gene Symbol CD200
Protein Tag His Tag
Validated Application Assay Standard
Protein Family CD Proteins
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 4345

Cluster of Differentiation 200 (CD200), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human CD200 at 1μg/mL (100μL/well) can bind human CD200R1-Fc with a linear range of 0.12-16ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.