missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD14 Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 345) of mouse CD14 precursor (NP_033971.1) fused to a polyhistidine tag at the C-terminus

$255.00 - $1,210.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias Cd14
Gene ID (Entrez) 12475
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50023M08H25 View Documents Life Technologies
LS50023M08H25
5 x 5μg Each for $255.00
 
50023M08HL View Documents Life Technologies
LS50023M08H250
5 x 50μg Each for $1,210.00
 
Description

Description

Cluster of Differentiation 14 (CD14), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
12475
<1 EU/μg
Cd14

For Research Use Only. Not for use in diagnostic procedures.