Please login to your online account to display your discounted pricing

Sino Biological™ CD14 Recombinant Human Protein, hIgG1-Fc Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the mature form of the extracellular domain (Thr 20-Cys 352) of human CD14 (NP_000582.1) fused to the Fc region of human IgG1 at the N-terminus

Manufacturer: life technologies  LS10073H01H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10073H01H25

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 62.4kDa
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Validated Application Assay Standard
Format Lyophilized
Protein Family CD Proteins
Gene Symbol CD14
Protein Tag IgG1 Fc Tag
Protein Subtype Glycoproteins
Quantity 5 x 5μg
Gene ID (Entrez) 929
Endotoxin Concentration <1 EU/μg
Structural Form Truncated/Recombinant/Cell Surface Protein
Gene Alias CD14 antigen, monocyte differentiation antigen CD14
Common Name CD14 molecule
Purity ≥90% by SDS-PAGE
Expression System Mammalian

Cluster of Differentiation 14 (CD14), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.