Please login to your online account to display your discounted pricing

Sino Biological™ CCL21 Recombinant Human Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human CCL21 (O00585) (Met 1-Pro 134)

$110.00 - $498.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias RP11-195F19.22-001, 6Ckine, CKb9, ECL, mgC34555, SCYA21, SLC, TCA4
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10477HNAB25

DFS Item
 
life technologies
LS10477HNAB25
5 x 5μg Each for $328.00

10477HNAB5

DFS Item
 
life technologies
LS10477HNAB5
5μg Each for $110.00

10477HNAB50

DFS Item
 
life technologies
LS10477HNAB50
50μg Each for $498.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias RP11-195F19.22-001, 6Ckine, CKb9, ECL, mgC34555, SCYA21, SLC, TCA4

Chemokine (C-C Motif) Ligand 21 (CCL21), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CCL21 at 2μg/mL (100μL/well) can bind human IGFBP7 with a linear range of 0.16-4μg/mL
  • Formulation: Lyophilized in 40mM Tris, 0.3M NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.