Please login to your online account to display your discounted pricing

Sino Biological™ CCL21 Recombinant Human Protein DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human CCL21 (O00585) (Met 1-Pro 134)

Manufacturer: life technologies  LS10477HNAB25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10477HNAB25

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 12.3kDa
Purity ≥90% by SDS-PAGE
Quantity 5 x 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Subtype Other Cytokines
Endotoxin Concentration <1 EU/μg
Gene Alias RP11-195F19.22-001, 6Ckine, CKb9, ECL, mgC34555, SCYA21, SLC, TCA4
Common Name Chemokine (C-C motif) ligand 21
Gene Symbol CCL21
Protein Tag Untagged
Validated Application Assay Standard
Protein Family Other Proteins
Expression System Insect
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized

Chemokine (C-C Motif) Ligand 21 (CCL21), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CCL21 at 2μg/mL (100μL/well) can bind human IGFBP7 with a linear range of 0.16-4μg/mL
  • Formulation: Lyophilized in 40mM Tris, 0.3M NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.