missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ CADM3 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human CADM3 (short isoform) (NP_001120645.1) extracellular domain (Met 1-His 330) fused to a polyhistidine tag at the C-terminus

$199.50 - $199.50

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CADM3, CTA-134P22.1, BIgR, FLJ10698, IGSF4B, NECL1, Necl-1, TSLL1, synCAM3
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
11214H08H25 Life Technologies
LS11214H08H25
5 x 5μg Each for $199.50
Specifications

Description

Cell Adhesion Molecule 3 (CADM3), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured by the ability of the immobilized protein to support the adhesion of C6 Rat brain glial cells; human CADM3 immobilized at 0.8μg/mL, 100μL/well will mediate >30% C6 cell adhesion
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Lyophilized
CADM3, CTA-134P22.1, BIgR, FLJ10698, IGSF4B, NECL1, Necl-1, TSLL1, synCAM3