Please login to your online account to display your discounted pricing

Sino Biological™ CA9 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human carbonic anhydrase 9 (CA9) precursor (NP_001207.2) (Met 1-Asp 414) fused to the polyhistidine tag at the C-terminus

$192.00 - $3,305.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CA9, CAIX, MN
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10107H08H25

DFS Item
 
life technologies
LS10107H08H25
5 x 5μg Each for $596.00

10107H08H5

DFS Item
 
life technologies
LS10107H08H5
5μg Each for $192.00

10107H08H50

DFS Item
 
life technologies
LS10107H08H50
50μg Each for $946.00

10107H08HL

DFS Item
 
life technologies
LS10107H08H250
5 x 50μg Each for $3,305.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CA9, CAIX, MN

Carbonic Anhydrase 9 (CA9), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its esterase activity; the specific activity is >30pmoles/min./μg, as measured with 1mM 4-Nitrophenyl acetate and 2.5μg enzyme at 400nM in 100μL of 12.5mM Tris, 75mM NaCl, pH 7.5
  • Formulation: Lyophilized in 25mM Tris, 150mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.