Please login to your online account to display your discounted pricing

Sino Biological™ CA9 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human carbonic anhydrase 9 (CA9) precursor (NP_001207.2) (Met 1-Asp 414) fused to the polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10107H08H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10107H08H25

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 48kDa
Purity ≥90% by SDS-PAGE
Format Lyophilized
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Gene Symbol CA9
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Protein Subtype Other Proteins
Quantity 5 x 5μg
Common Name Carbonic anhydrase IX
Gene Alias CA9, CAIX, MN
Protein Family Other Proteins
Expression System Mammalian
Endotoxin Concentration <1 EU/μg
Protein Tag His Tag
Validated Application Assay Standard

Carbonic Anhydrase 9 (CA9), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its esterase activity; the specific activity is >30pmoles/min./μg, as measured with 1mM 4-Nitrophenyl acetate and 2.5μg enzyme at 400nM in 100μL of 12.5mM Tris, 75mM NaCl, pH 7.5
  • Formulation: Lyophilized in 25mM Tris, 150mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.